Assay Details
| Gene Symbol: Eya1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001252192.1 | NP_001239121.1 | 7 - 8 | 1120 | 59 | |
| NM_010164.2 | NP_034294.2 | 8 - 9 | 1236 | 59 | ||
| GenBank mRNA | AK042412.1 | - | 7 - 8 | 1043 | 59 | |
| BC060260.1 | - | 8 - 9 | 1236 | 6848408 | 59 | |
| BC066860.1 | - | 8 - 9 | 890 | 30089692 | 59 | |
| U61110.1 | - | 7 - 8 | 793 | 59 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Eya1
Genomic Map