Assay Details
| Gene Symbol: Rbm34 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172762.2 | NP_766350.2 | 5 - 6 | 711 | 151 | |
| GenBank mRNA | AK078099.1 | - | 5 - 6 | 704 | 151 | |
| BC064035.1 | - | 5 - 6 | 690 | 151 | ||
| BC141278.1 | - | 5 - 6 | 700 | 9055890 | 151 | |
More Information
Updated Mapping Information
Set Membership: |
|

Rbm34
Genomic Map