This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01239826_g1
See other RBM34 GE Assays ›
Gene Symbol:
Rbm34
Gene Name:
RNA binding motif protein 34
Gene Aliases:
4930547K05Rik, 6330444B11, D8Ertd233e
Chromosome Location:
Chr.8: 126947172 - 126971126 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01239826_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Rbm34
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_172762.2 NP_766350.2 7 - 8 833 67
GenBank mRNA AK016057.1 - 6 - 7 765 67
AK078099.1 - 7 - 8 826 67
BC064035.1 - 7 - 8 812 67
BC141278.1 - 7 - 8 822 9055890 67

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0800003202.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Rbm34

Panther Classification:

Molecular Function -

translation initiation factor

Gene Ontology Categories:

Function(s) Process(es)

biological_process
nucleotide binding
nucleic acid binding
RNA binding
poly(A) RNA binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0800024631.mm.1 NM_172762.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0800024631.mm.1 NM_172762.2 Refseq
Affymetrix Mouse Clariom S Array PSR0800024631.mm.1 TC0800003202.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-x4xlz:80/100.66.78.26:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0