This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01240153_g1
See other ASXL1 GE Assays ›
Gene Symbol:
Asxl1
Gene Name:
additional sex combs like 1
Gene Aliases:
mKIAA0978
Chromosome Location:
Chr.2: 153346139 - 153404007 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01240153_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Asxl1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001039939.1 NP_001035028.1 11 - 12 1277 80
GenBank mRNA AK122413.1 - 11 - 12 1277 80

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0200002315.mm.1

Set Membership:

Amplicon length between 71 and 85 Detect Genomic DNA Mus musculus Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

negative regulation of transcription from RNA polymerase II promoter
heart morphogenesis
transcription, DNA-templated
regulation of transcription, DNA-templated
chromatin modification
response to retinoic acid
negative regulation of peroxisome proliferator activated receptor signaling pathway
monoubiquitinated histone H2A deubiquitination
negative regulation of fat cell differentiation
positive regulation of transcription from RNA polymerase II promoter
positive regulation of retinoic acid receptor signaling pathway
negative regulation of retinoic acid receptor signaling pathway
bone development
lung saccule development
DNA binding
transcription coactivator activity
transcription corepressor activity
protein binding
retinoic acid receptor binding
peroxisome proliferator activated receptor binding
metal ion binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0200018223.mm.1 NM_001039939.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200018223.mm.1 NM_001039939.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200018223.mm.1 TC0200002315.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-pkm9c:80/100.66.79.31:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0