This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01240348_m1
See other MAPT GE Assays ›
Gene Symbol:
Mapt
Gene Name:
microtubule-associated protein tau
Gene Aliases:
AI413597, AW045860, Mtapt, Tau
Chromosome Location:
Chr.11: 104231353 - 104332090 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01240348_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Mapt
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001038609.2 NP_001033698.1 1 - 2 168 58
NM_001285454.1 NP_001272383.1 1 - 2 168 58
NM_001285455.1 NP_001272384.1 1 - 2 168 58
NM_001285456.1 NP_001272385.1 1 - 2 168 58
NM_010838.4 NP_034968.3 1 - 2 168 58
GenBank mRNA AK038797.1 - 1 - 2 125 58
AK078167.1 - 1 - 2 142 58
AK147636.1 - 1 - 2 129 58
BC014748.1 - 1 - 2 106 4504850 58
BI202659.1 - 1 - 2 58
BY249380.1 - 1 - 2 163 58
CO432693.1 - 1 - 2 118 58
M18775.1 - 1 - 2 169 58
M18776.1 - 1 - 2 169 58
W18770.1 - 1 - 2 150 58

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the AK078167.1, AK147636.1, BC014748.1, M18775.1, M18776.1, NM_001038609.2, NM_001285454.1, NM_001285455.1, NM_001285456.1, NM_010838.4 transcripts.

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Apoptosis Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Mapt

Gene Ontology Categories:

Function(s) Process(es)

microtubule cytoskeleton organization
neuron migration
apoptotic process
axonogenesis
response to nutrient
adult walking behavior
axo-dendritic transport
intrinsic apoptotic signaling pathway in response to oxidative stress
response to organic substance
regulation of autophagy
positive regulation of microtubule polymerization
negative regulation of intracellular transport
positive regulation of axon extension
mitochondrion transport along microtubule
axon extension
regulation of microtubule-based movement
protein binding
microtubule binding
tubulin binding
SH3 domain binding
enzyme binding
protein kinase binding
protein domain specific binding
protein complex binding
apolipoprotein binding
protein phosphatase 2A binding
lipoprotein particle binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1100016288.mm.1 NM_001038609.2 Refseq
Affymetrix Mouse Clariom S Array PSR1100016288.mm.1 NM_001285454.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100016288.mm.1 NM_001285455.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100016288.mm.1 NM_001285456.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100016288.mm.1 NM_010838.4 Refseq
Affymetrix Mouse Clariom S Array PSR1100016288.mm.1 TC1100001725.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1100016298.mm.1 NM_001038609.2 Refseq
Affymetrix Mouse Clariom S Array PSR1100016298.mm.1 NM_001285454.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100016298.mm.1 NM_001285455.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100016298.mm.1 NM_001285456.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100016298.mm.1 NM_010838.4 Refseq
Affymetrix Mouse Clariom S Array PSR1100016298.mm.1 TC1100001725.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1100016292.mm.1 NM_001038609.2 Refseq
Affymetrix Mouse Clariom S Array PSR1100016292.mm.1 NM_001285454.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100016292.mm.1 NM_001285455.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100016292.mm.1 NM_001285456.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100016292.mm.1 NM_010838.4 Refseq
Affymetrix Mouse Clariom S Array PSR1100016292.mm.1 TC1100001725.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-f8k5r:80/100.66.76.16:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline