This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01240505_m1
See other PITPNA GE Assays ›
Gene Symbol:
Pitpna
Gene Name:
phosphatidylinositol transfer protein, alpha
Gene Aliases:
Pitpn, vb, vibrator
Chromosome Location:
Chr.11: 75588108 - 75628778 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01240505_m1
Size
Availability Made To Order
Catalog # 4351372
Price 417.00
Your Price
Online offer: 387.65
387.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Pitpna
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_008850.1 NP_032876.1 9 - 10 990 67
GenBank mRNA AK144618.1 - 10 - 11 1003 67
AK149745.1 - 10 - 11 980 67
AK151496.1 - 10 - 11 979 67
AK152089.1 - 10 - 11 1003 67
AK152874.1 - 10 - 11 997 67
AK153569.1 - 10 - 11 1003 67
AK165659.1 - 10 - 11 995 67
AK167760.1 - 9 - 10 1003 67
AK168187.1 - 9 - 10 960 67
AK168720.1 - 10 - 11 993 67
AK186336.1 - 181 67
AK187601.1 - 3 - 4 253 67
BC056171.1 - 10 - 11 945 5710926 67
S72681.1 - 9 - 10 765 67
U96725.1 - 9 - 10 991 67

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1100001074.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Transporter Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Pitpna

Panther Classification:

Molecular Function -

transporter

Gene Ontology Categories:

Function(s) Process(es)

transport
phospholipid transport
fatty-acyl-CoA binding
phospholipid binding
lipid binding
phosphatidylinositol transporter activity
phosphatidylinositol binding
stearic acid binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1100009791.mm.1 NM_008850.2 Refseq
Affymetrix Mouse Clariom S Array PSR1100009791.mm.1 TC1100001074.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-blq6p:80/100.66.74.192:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline