This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01240572_g1
See other PLTP GE Assays ›
Gene Symbol:
Pltp
Gene Name:
phospholipid transfer protein
Gene Aliases:
Bpife, OD107
Chromosome Location:
Chr.2: 164839518 - 164857708 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01240572_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Pltp
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_011125.2 NP_035255.1 2 - 3 240 85
GenBank mRNA AK148330.1 - 2 - 3 245 85
AK149730.1 - 2 - 3 171 85
U28960.1 - 1 - 2 98 85
U37226.1 - 2 - 3 170 85

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Pltp

Gene Ontology Categories:

Function(s) Process(es)

lipid metabolic process
transport
lipid transport
vitamin E biosynthetic process
positive regulation of cholesterol efflux
sperm motility
high-density lipoprotein particle remodeling
lipid binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-9dqhd:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline