This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01240765_m1
See other TCF15 GE Assays ›
Gene Symbol:
Tcf15
Gene Name:
transcription factor 15
Gene Aliases:
Meso1, bHLH-EC2
Chromosome Location:
Chr.2: 152143046 - 152149097 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01240765_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Tcf15
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_009328.2 NP_033354.2 1 - 2 533 80
GenBank mRNA BC027533.1 - 1 - 2 533 5143248 80
U18658.1 - 1 - 2 573 80

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Tcf15

Panther Classification:

Molecular Function -

basic helix-loop-helix transcription factor

Gene Ontology Categories:

Function(s) Process(es)

somitogenesis
respiratory system process
transcription, DNA-templated
regulation of transcription, DNA-templated
multicellular organism development
muscle organ development
anterior/posterior pattern specification
regulation of gene expression
obsolete death
post-anal tail morphogenesis
eating behavior
ear development
skin development
establishment of epithelial cell apical/basal polarity
paraxial mesoderm development
muscle organ morphogenesis
skeletal system morphogenesis
neuromuscular process controlling posture
mesenchymal to epithelial transition
regulation of extracellular matrix organization
DNA binding
protein dimerization activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-gwzxr:80/100.66.76.73:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0