This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01240880_m1
See other MBTPS1 GE Assays ›
Gene Symbol:
Mbtps1
Gene Name:
membrane-bound transcription factor peptidase, site 1
Gene Aliases:
0610038M03Rik, AV003995, S1P, SKI-1, Ski1, mKIAA0091
Chromosome Location:
Chr.8: 119508151 - 119558825 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01240880_m1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Mbtps1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001167910.1 NP_001161382.1 17 - 18 2789 91
NM_019709.4 NP_062683.3 18 - 19 2938 91
GenBank mRNA AF094820.1 - 16 - 17 2357 91
AK029048.1 - 17 - 18 2766 91
AK122202.1 - 17 - 18 2755 91
AK147336.1 - 16 - 17 5358 91
AK147495.1 - 17 - 18 2763 91
BC011533.1 - 17 - 18 2752 91
BC054837.1 - 16 - 17 2557 6490394 91
BC057198.1 - 18 - 19 2904 5310414 91
DQ913734.1 - 16 - 17 2420 91

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Mbtps1

Panther Classification:

Molecular Function -

serine protease

Gene Ontology Categories:

Function(s) Process(es)

proteolysis
lipid metabolic process
lysosome organization
steroid metabolic process
cholesterol metabolic process
regulation of transcription factor import into nucleus
endopeptidase activity
serine-type endopeptidase activity
peptidase activity
serine-type peptidase activity
hydrolase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-744bc48644-fvx7n:80/100.66.78.247:80.
git-commit: 747bde55a712f6f97bf7760408d445eefba4e16f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.48.0-Offline