Assay Details
| Gene Symbol: Flot2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001284228.1 | NP_001271157.1 | 3 - 4 | 205 | 63 | |
| NM_008028.2 | NP_032054.1 | 3 - 4 | 349 | 63 | ||
| GenBank mRNA | BC070423.1 | - | 3 - 4 | 307 | 6403816 | 63 |
| U07890.1 | - | 3 - 4 | 219 | 63 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the BC070423.1, NM_001284228.1, NM_008028.2, U07890.1 transcripts.
Set Membership: |
|

Flot2
Genomic Map