This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01241588_g1
See other SLC7A5 GE Assays ›
Gene Symbol:
Slc7a5
Gene Name:
solute carrier family 7 (cationic amino acid transporter, y+ system), member 5
Gene Aliases:
4F2LC, D0H16S474E, LAT1, TA1
Chromosome Location:
Chr.8: 121881146 - 121907686 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01241588_g1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer:
435.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Slc7a5
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_011404.3 NP_035534.2 6 - 7 1133 92
GenBank mRNA AB017189.1 - 6 - 7 1079 92
AB023409.1 - 6 - 7 1061 92
AK142288.1 - 6 - 7 1134 92
AK148134.1 - 6 - 7 803 92
BC026131.1 - 6 - 7 1142 4188854 92

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0800003059.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Transporter Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Slc7a5

Panther Classification:

Molecular Function -

transporter

Gene Ontology Categories:

Function(s) Process(es)

amino acid transmembrane transport
transport
amino acid transport
multicellular organism development
nervous system development
L-amino acid transport
leucine transport
phenylalanine transport
tryptophan transport
cell differentiation
cation transmembrane transport
L-alpha-amino acid transmembrane transport
amino acid transmembrane transporter activity
L-amino acid transmembrane transporter activity
L-leucine transmembrane transporter activity
L-phenylalanine transmembrane transporter activity
L-tryptophan transmembrane transporter activity
antiporter activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-zvjvc:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline