This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01241959_g1
See other GAA GE Assays ›
Gene Symbol:
Gaa
Gene Name:
glucosidase, alpha, acid
Gene Aliases:
E430018M07Rik
Chromosome Location:
Chr.11: 119267967 - 119285698 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01241959_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Gaa
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001159324.1 NP_001152796.1 16 - 17 2500 59
NM_008064.3 NP_032090.3 16 - 17 2590 59
GenBank mRNA AK052211.1 - 16 - 17 2571 59
AK088481.1 - 16 - 17 2566 59
AK139333.1 - 16 - 17 2538 59
AK141546.1 - 16 - 17 2573 59
AK145631.1 - 15 - 16 2006 59
AK146538.1 - 16 - 17 2494 59
AK150970.1 - 16 - 17 2548 59

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Gaa

Panther Classification:

Molecular Function -

glucosidase

Gene Ontology Categories:

Function(s) Process(es)

regulation of the force of heart contraction
diaphragm contraction
heart morphogenesis
carbohydrate metabolic process
glycogen metabolic process
glycogen catabolic process
striated muscle contraction
lysosome organization
locomotory behavior
metabolic process
tissue development
vacuolar sequestering
muscle cell cellular homeostasis
neuromuscular process controlling posture
neuromuscular process controlling balance
cardiac muscle contraction
catalytic activity
hydrolase activity, hydrolyzing O-glycosyl compounds
alpha-1,4-glucosidase activity
hydrolase activity
hydrolase activity, acting on glycosyl bonds
carbohydrate binding
maltose alpha-glucosidase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-flftk:80/100.66.79.227:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0