This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01242095_m1
See other CDH8 GE Assays ›
Gene Symbol:
Cdh8
Gene Name:
cadherin 8
Gene Aliases:
AI851472, cad8
Chromosome Location:
Chr.8: 99024471 - 99416485 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01242095_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Cdh8
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001039154.2 NP_001034243.1 5 - 6 1420 89
NM_001285913.1 NP_001272842.1 5 - 6 1420 89
NM_001285914.1 NP_001272843.1 5 - 6 1420 89
NM_007667.3 NP_031693.2 5 - 6 1420 89
GenBank mRNA AK044046.1 - 5 - 6 1378 89
AK047589.1 - 3 - 4 351 89
AK083092.1 - 5 - 6 1387 89
AK086711.1 - 5 - 6 1389 89
AK139334.1 - 5 - 6 1376 89
BC057581.1 - 5 - 6 1325 6416918 89
BC060200.1 - 5 - 6 1268 89
X95600.1 - 5 - 6 1060 89

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0800002767.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Cdh8

Panther Classification:

Molecular Function -

cadherin

Gene Ontology Categories:

Function(s) Process(es)

cell adhesion
homophilic cell adhesion via plasma membrane adhesion molecules
chemical synaptic transmission
response to cold
synaptic transmission, glutamatergic
calcium ion binding
metal ion binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0800021279.mm.1 NM_001039154.2 Refseq
Affymetrix Mouse Clariom S Array PSR0800021279.mm.1 NM_001285913.1 Refseq
Affymetrix Mouse Clariom S Array PSR0800021279.mm.1 NM_001285914.1 Refseq
Affymetrix Mouse Clariom S Array PSR0800021279.mm.1 NM_007667.3 Refseq
Affymetrix Mouse Clariom S Array PSR0800021279.mm.1 TC0800002767.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-zpsr4:80/100.66.78.116:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0