This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01242096_m1
See other CDH8 GE Assays ›
Gene Symbol:
Cdh8
Gene Name:
cadherin 8
Gene Aliases:
AI851472, cad8
Chromosome Location:
Chr.8: 99024471 - 99416485 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01242096_m1
Size
Availability Inventoried
Catalog # 4331182
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Cdh8
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001039154.2 NP_001034243.1 6 - 7 1614 78
NM_001285913.1 NP_001272842.1 6 - 7 1614 78
NM_001285914.1 NP_001272843.1 6 - 7 1614 78
NM_007667.3 NP_031693.2 6 - 7 1614 78
GenBank mRNA AK044046.1 - 6 - 7 1572 78
AK047589.1 - 4 - 5 545 78
AK083092.1 - 6 - 7 1581 78
AK086711.1 - 6 - 7 1583 78
AK139334.1 - 6 - 7 1570 78
BC057581.1 - 6 - 7 1519 6416918 78
BC060200.1 - 6 - 7 1462 78
X95600.1 - 6 - 7 1254 78

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0800002767.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus 1700 Inventoried Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Cdh8

Panther Classification:

Molecular Function -

cadherin

Gene Ontology Categories:

Function(s) Process(es)

cell adhesion
homophilic cell adhesion via plasma membrane adhesion molecules
chemical synaptic transmission
response to cold
synaptic transmission, glutamatergic
calcium ion binding
metal ion binding

MicroArray Probes:

Affymetrix ABI

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0800021278.mm.1 NM_001039154.2 Refseq
Affymetrix Mouse Clariom S Array PSR0800021278.mm.1 NM_001285913.1 Refseq
Affymetrix Mouse Clariom S Array PSR0800021278.mm.1 NM_001285914.1 Refseq
Affymetrix Mouse Clariom S Array PSR0800021278.mm.1 NM_007667.3 Refseq
Affymetrix Mouse Clariom S Array PSR0800021278.mm.1 TC0800002767.mm.1 MTA TC
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 463713 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-jbr9k:80/100.66.74.50:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0