This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01242359_g1
See other P2RX5 GE Assays ›
Gene Symbol:
P2rx5
Gene Name:
purinergic receptor P2X, ligand-gated ion channel, 5
Gene Aliases:
P2X5
Chromosome Location:
Chr.11: 73155277 - 73175327 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01242359_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: P2rx5
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_033321.3 NP_201578.2 6 - 7 685 87
GenBank mRNA AF333331.1 - 6 - 7 691 87
AK134662.1 - 6 - 7 645 87
BC138189.1 - 6 - 7 685 8861208 87
BC145181.1 - 6 - 7 685 9053732 87

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Ion Channel Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene P2rx5

Panther Classification:

Molecular Function -

ligand-gated ion channel

Gene Ontology Categories:

Function(s) Process(es)

cell surface receptor signaling pathway
neuronal action potential
purinergic nucleotide receptor signaling pathway
regulation of skeletal muscle tissue regeneration
protein homooligomerization
cation transmembrane transport
mitophagy in response to mitochondrial depolarization
purinergic nucleotide receptor activity
extracellular ATP-gated cation channel activity
ATP binding
GTP binding
drug binding
zinc ion binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1100009216.mm.1 NM_033321.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100009216.mm.1 NM_033321.3 Refseq
Affymetrix Mouse Clariom S Array PSR1100009216.mm.1 TC1100001016.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-x88k6:80/100.66.75.98:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0