This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01242426_m1
See other CNTNAP4 GE Assays ›
Gene Symbol:
Cntnap4
Gene Name:
contactin associated protein-like 4
Gene Aliases:
Caspr4, E130114F09Rik
Chromosome Location:
Chr.8: 112569965 - 112882707 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01242426_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Cntnap4
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_130457.2 NP_569724.2 4 - 5 637 88
GenBank mRNA AF333770.1 - 4 - 5 602 88
AK042607.1 - 4 - 5 626 88
AK053611.1 - 4 - 5 629 88
AK081891.1 - 4 - 5 630 88
AK142816.1 - 4 - 5 633 88
AK173255.1 - 4 - 5 612 88
BC034628.1 - 4 - 5 637 5369648 88

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Cntnap4

Gene Ontology Categories:

Function(s) Process(es)

cell adhesion
regulation of synaptic transmission, dopaminergic
regulation of synaptic transmission, GABAergic
regulation of grooming behavior
protein binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-8cb8z:80/100.66.72.77:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0