This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01242701_m1
See other MILR1 GE Assays ›
Gene Symbol:
Milr1
Gene Name:
mast cell immunoglobulin like receptor 1
Gene Aliases:
Allergin-1, Gm885, Mca32
Chromosome Location:
Chr.11: 106750652 - 106768794 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01242701_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Milr1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001033435.3 NP_001028607.1 3 - 4 478 105
GenBank mRNA AB542953.1 - 3 - 4 436 105
AK136488.1 - 3 - 4 438 105
AK151814.1 - 3 - 4 436 105
AK153506.1 - 3 - 4 386 105
AK170947.1 - 3 - 4 437 105
BC139224.1 - 3 - 4 464 8862246 105
BC139225.1 - 3 - 4 464 8862247 105

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1100001785.mm.1

Set Membership:

Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons

Panther Classification:

Molecular Function -

immunoglobulin receptor superfamily

Gene Ontology Categories:

Function(s) Process(es)

negative regulation of mast cell activation
mast cell degranulation
protein binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1100016856.mm.1 NM_001033435.3 Refseq
Affymetrix Mouse Clariom S Array PSR1100016856.mm.1 NM_001271372.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100016856.mm.1 NM_001271373.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100016856.mm.1 NM_001271374.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100016856.mm.1 NM_001271375.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100016856.mm.1 NR_073165.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100016856.mm.1 TC1100001785.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-wf7gw:80/100.66.73.223:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0