This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01242771_g1
See other SMARCD2 GE Assays ›
Gene Symbol:
Smarcd2
Gene Name:
SWI/SNF related, matrix associated, actin dependent regulator of chromatin, subfamily d, member 2
Gene Aliases:
AW322457, Baf60b
Chromosome Location:
Chr.11: 106263171 - 106272972 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01242771_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Smarcd2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001130187.1 NP_001123659.1 5 - 6 983 98
NM_031878.2 NP_114084.2 4 - 5 617 98
GenBank mRNA AK159259.1 - 5 - 6 820 98
AK166123.1 - 5 - 6 777 98
AK216602.1 - 1 - 2 55 98
BC005732.1 - 4 - 5 607 2647494 98

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the AK166123.1 transcripts.

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Smarcd2

Panther Classification:

Molecular Function -

chromatin/chromatin-binding, or -regulatory protein

Gene Ontology Categories:

Function(s) Process(es)

nucleosome disassembly
chromatin remodeling
transcription, DNA-templated
regulation of transcription, DNA-templated
chromatin modification
ATP-dependent chromatin remodeling
RNA polymerase II core promoter proximal region sequence-specific DNA binding
RNA polymerase II distal enhancer sequence-specific DNA binding
nucleosomal DNA binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1100036694.mm.1 NM_001130187.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100036694.mm.1 NM_001130187.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100036694.mm.1 NM_031878.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100036694.mm.1 NM_031878.2 Refseq
Affymetrix Mouse Clariom S Array PSR1100036694.mm.1 TC1100003918.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-f8k5r:80/100.66.76.16:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline