This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01243003_g1
See other GNPAT GE Assays ›
Gene Symbol:
Gnpat
Gene Name:
glyceronephosphate O-acyltransferase
Gene Aliases:
AU019525, D1Ertd819e, DHAPAT
Chromosome Location:
Chr.8: 124863033 - 124890057 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01243003_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Gnpat
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_010322.3 NP_034452.3 13 - 14 2043 67
GenBank mRNA AF110769.1 - 13 - 14 2007 67
AJ132012.1 - 13 - 14 1838 67
AK004364.1 - 4 - 5 490 67
AK004630.1 - 13 - 14 2017 67
AK004771.1 - 13 - 14 2023 67
AK010896.1 - 13 - 14 2017 67
AK150810.1 - 13 - 14 2001 67
AK208037.1 - 3 - 4 290 67
BC025972.1 - 13 - 14 2000 4020505 67

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0800003239.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Gnpat

Panther Classification:

Molecular Function -

acyltransferase

Gene Ontology Categories:

Function(s) Process(es)

synapse assembly
metabolic process
ether lipid biosynthetic process
cerebellum morphogenesis
paranodal junction assembly
myelination
cellular lipid metabolic process
membrane organization
catalytic activity
receptor binding
protein binding
O-acyltransferase activity
glycerone-phosphate O-acyltransferase activity
palmitoyl-CoA hydrolase activity
transferase activity
transferase activity, transferring acyl groups

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0800012748.mm.1 NM_010322.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0800012748.mm.1 NM_010322.3 Refseq
Affymetrix Mouse Clariom S Array PSR0800012748.mm.1 TC0800003239.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0800012746.mm.1 NM_010322.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0800012746.mm.1 NM_010322.3 Refseq
Affymetrix Mouse Clariom S Array PSR0800012746.mm.1 TC0800003239.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-sc64w:80/100.66.79.87:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0