Assay Details
| Gene Symbol: Lrrc45 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_153545.2 | NP_705773.2 | 14 - 15 | 1531 | 78 | |
| GenBank mRNA | AK081566.1 | - | 13 - 14 | 1459 | 78 | |
| AK153361.1 | - | 14 - 15 | 1532 | 78 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Lrrc45
Genomic Map