This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01243452_g1
See other GOT2 GE Assays ›
Gene Symbol:
Got2
Gene Name:
glutamatic-oxaloacetic transaminase 2, mitochondrial
Gene Aliases:
AL022787, FABP-pm, Got-2, mAspAT
Chromosome Location:
Chr.8: 95864137 - 95888365 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01243452_g1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Got2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_010325.2 NP_034455.1 5 - 6 659 66
GenBank mRNA AK098166.1 - 5 - 6 685 66
AK136556.1 - 5 - 6 696 66
AK147953.1 - 5 - 6 699 66
AK149886.1 - 5 - 6 696 66
AK149926.1 - 5 - 6 696 66
AK150194.1 - 4 - 5 415 66
AK152921.1 - 5 - 6 696 66
AK155075.1 - 5 - 6 690 66
AK167767.1 - 5 - 6 685 66
AK190359.1 - 1 - 2 39 66
AK219323.1 - 1 - 2 39 66
BC089015.1 - 5 - 6 685 30606379 66
BC089341.1 - 5 - 6 697 6832287 66
U82470.1 - 5 - 6 627 66

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Got2

Panther Classification:

Molecular Function -

transaminase

Gene Ontology Categories:

Function(s) Process(es)

oxaloacetate metabolic process
cellular amino acid metabolic process
aspartate metabolic process
aspartate biosynthetic process
aspartate catabolic process
transport
lipid transport
biosynthetic process
glutamate catabolic process to aspartate
glutamate catabolic process to 2-oxoglutarate
dicarboxylic acid metabolic process
response to ethanol
catalytic activity
L-aspartate:2-oxoglutarate aminotransferase activity
phospholipid binding
transaminase activity
kynurenine-oxoglutarate transaminase activity
amino acid binding
transferase activity
enzyme binding
pyridoxal phosphate binding
carboxylic acid binding
protein homodimerization activity
poly(A) RNA binding
L-phenylalanine:2-oxoglutarate aminotransferase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0800021240.mm.1 NM_010325.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0800021240.mm.1 NM_010325.2 Refseq
Affymetrix Mouse Clariom S Array PSR0800021240.mm.1 TC0800002758.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-744bc48644-454p7:80/100.66.72.101:80.
git-commit: 747bde55a712f6f97bf7760408d445eefba4e16f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.48.0-Offline