This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01243531_m1
See other PDPR GE Assays ›
Gene Symbol:
Pdpr
Gene Name:
pyruvate dehydrogenase phosphatase regulatory subunit
Gene Aliases:
4930402E16Rik, AU018056, mKIAA1990
Chromosome Location:
Chr.8: 111094599 - 111139779 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01243531_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Pdpr
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_198308.1 NP_938050.1 5 - 6 739 66
GenBank mRNA AK029575.1 - 5 - 6 830 66
AK129472.1 - 5 - 6 843 66
AY223867.1 - 5 - 6 739 66
BC120745.1 - 4 - 5 669 40129668 66
BC125425.1 - 4 - 5 669 40129940 66

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0800001328.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Pdpr

Panther Classification:

Molecular Function -

methyltransferase

Gene Ontology Categories:

Function(s) Process(es)

oxidation-reduction process
molecular_function
oxidoreductase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0800010936.mm.1 NM_198308.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0800010936.mm.1 NM_198308.1 Refseq
Affymetrix Mouse Clariom S Array PSR0800010936.mm.1 TC0800001328.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-wf7gw:80/100.66.73.223:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0