This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01243748_m1
See other TOM1 GE Assays ›
Gene Symbol:
Tom1
Gene Name:
target of myb1 homolog (chicken)
Gene Aliases:
-
Chromosome Location:
Chr.8: 75033686 - 75070121 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01243748_m1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Tom1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001136259.1 NP_001129731.1 1 - 2 143 87
NM_011622.3 NP_035752.1 1 - 2 143 87
GenBank mRNA AJ006972.1 - 1 - 2 120 87
AK028398.1 - 1 - 2 84 87
AK051245.1 - 1 - 2 125 87
AK150143.1 - 1 - 2 93 87
AK160403.1 - 1 - 2 89 87
BC021633.1 - 1 - 2 106 4208226 87
BC093520.1 - 1 - 2 71 2866013 87
BC157905.1 - 1 - 2 104 9088106 87
BC158042.1 - 1 - 2 104 9007343 87

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the AJ006972.1, AK028398.1, AK051245.1, AK150143.1, AK160403.1, BC021633.1, BC093520.1, BC157905.1, BC158042.1, NM_001136259.1, NM_011622.3 transcripts.

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Transporter Mammalian Gene Collection Mus musculus 1700 Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Tom1

Gene Ontology Categories:

Function(s) Process(es)

transport
intracellular protein transport
protein transport
protein binding
clathrin binding

MicroArray Probes:

Affymetrix ABI

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0800006241.mm.1 NM_001136259.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0800006241.mm.1 NM_001136259.1 Refseq
Affymetrix Mouse Clariom S Array PSR0800006241.mm.1 NM_011622.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0800006241.mm.1 NM_011622.3 Refseq
Affymetrix Mouse Clariom S Array PSR0800006241.mm.1 TC0800000844.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0800006242.mm.1 NM_001136259.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0800006242.mm.1 NM_001136259.1 Refseq
Affymetrix Mouse Clariom S Array PSR0800006242.mm.1 NM_011622.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0800006242.mm.1 NM_011622.3 Refseq
Affymetrix Mouse Clariom S Array PSR0800006242.mm.1 TC0800000844.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0800006238.mm.1 NM_001136259.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0800006238.mm.1 NM_001136259.1 Refseq
Affymetrix Mouse Clariom S Array PSR0800006238.mm.1 NM_011622.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0800006238.mm.1 NM_011622.3 Refseq
Affymetrix Mouse Clariom S Array PSR0800006238.mm.1 TC0800000844.mm.1 MTA TC
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 620405 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-d7bxm:80/100.66.76.204:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0