This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01243749_g1
See other TOM1 GE Assays ›
Gene Symbol:
Tom1
Gene Name:
target of myb1 homolog (chicken)
Gene Aliases:
-
Chromosome Location:
Chr.8: 75033686 - 75070121 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01243749_g1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Tom1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001136259.1 NP_001129731.1 4 - 5 461 92
NM_011622.3 NP_035752.1 4 - 5 461 92
GenBank mRNA AJ006972.1 - 4 - 5 438 92
AK028398.1 - 4 - 5 402 92
AK051245.1 - 4 - 5 443 92
AK150143.1 - 4 - 5 411 92
AK160403.1 - 4 - 5 407 92
BC021633.1 - 4 - 5 424 4208226 92
BC093520.1 - 4 - 5 389 2866013 92
BC157905.1 - 4 - 5 422 9088106 92
BC158042.1 - 4 - 5 422 9007343 92

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Transporter Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Tom1

Gene Ontology Categories:

Function(s) Process(es)

transport
intracellular protein transport
protein transport
protein binding
clathrin binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0800006245.mm.1 NM_001136259.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0800006245.mm.1 NM_001136259.1 Refseq
Affymetrix Mouse Clariom S Array PSR0800006245.mm.1 NM_011622.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0800006245.mm.1 NM_011622.3 Refseq
Affymetrix Mouse Clariom S Array PSR0800006245.mm.1 TC0800000844.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-vh84l:80/100.66.79.51:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0