This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01243751_g1
See other TOM1 GE Assays ›
Gene Symbol:
Tom1
Gene Name:
target of myb1 homolog (chicken)
Gene Aliases:
-
Chromosome Location:
Chr.8: 75033686 - 75070121 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01243751_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Tom1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001136259.1 NP_001129731.1 9 - 10 1019 78
NM_011622.3 NP_035752.1 9 - 10 1019 78
GenBank mRNA AJ006972.1 - 9 - 10 996 78
AK028398.1 - 9 - 10 960 78
AK051245.1 - 9 - 10 1001 78
AK150143.1 - 9 - 10 969 78
AK160403.1 - 9 - 10 965 78
AK189274.1 - 2 - 3 51 78
BC021633.1 - 9 - 10 982 4208226 78
BC093520.1 - 9 - 10 947 2866013 78
BC157905.1 - 9 - 10 980 9088106 78
BC158042.1 - 9 - 10 980 9007343 78

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Transporter Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Tom1

Gene Ontology Categories:

Function(s) Process(es)

transport
intracellular protein transport
protein transport
protein binding
clathrin binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0800006250.mm.1 NM_001136259.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0800006250.mm.1 NM_001136259.1 Refseq
Affymetrix Mouse Clariom S Array PSR0800006250.mm.1 NM_011622.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0800006250.mm.1 NM_011622.3 Refseq
Affymetrix Mouse Clariom S Array PSR0800006250.mm.1 TC0800000844.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0800006251.mm.1 NM_001136259.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0800006251.mm.1 NM_001136259.1 Refseq
Affymetrix Mouse Clariom S Array PSR0800006251.mm.1 TC0800000844.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-mvcqg:80/100.66.75.199:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0