This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01244208_g1
See other LDHD GE Assays ›
Gene Symbol:
Ldhd
Gene Name:
lactate dehydrogenase D
Gene Aliases:
4733401P21Rik, D8Bwg1320e
Chromosome Location:
Chr.8: 111626271 - 111630322 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01244208_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Ldhd
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_027570.3 NP_081846.3 6 - 7 840 83
GenBank mRNA AK014649.1 - 6 - 7 846 83
AK029820.1 - 1254 83
BC039155.1 - 6 - 7 836 4242682 83
BC055443.1 - 6 - 7 840 4235345 83

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0800002950.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Ldhd

Panther Classification:

Molecular Function -

dehydrogenase

Gene Ontology Categories:

Function(s) Process(es)

ATP biosynthetic process
oxidation-reduction process
catalytic activity
D-lactate dehydrogenase (cytochrome) activity
protein binding
D-lactate dehydrogenase activity
UDP-N-acetylmuramate dehydrogenase activity
oxidoreductase activity
oxidoreductase activity, acting on CH-OH group of donors
flavin adenine dinucleotide binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0800022790.mm.1 NM_027570.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0800022790.mm.1 NM_027570.3 Refseq
Affymetrix Mouse Clariom S Array PSR0800022790.mm.1 TC0800002950.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0800022792.mm.1 NM_027570.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0800022792.mm.1 NM_027570.3 Refseq
Affymetrix Mouse Clariom S Array PSR0800022792.mm.1 TC0800002950.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-55658555d6-qfdf2:80/100.66.76.55:80.
git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.41.0-Offline