This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01244860_m1
See other UCP1 GE Assays ›
Gene Symbol:
Ucp1
Gene Name:
uncoupling protein 1 (mitochondrial, proton carrier)
Gene Aliases:
AI385626, Slc25a7, Ucp
Chromosome Location:
Chr.8: 83290348 - 83298456 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01244860_m1
Size
Availability Inventoried
Catalog # 4331182
Price 2,694.00
Your Price
Online offer:
2694.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information
  • Citations (1)

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Ucp1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_009463.3 NP_033489.1 4 - 5 862 140
GenBank mRNA AK002759.1 - 4 - 5 860 140
BC012701.1 - 4 - 5 859 4191276 140
U63419.1 - 4 - 5 858 140

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0800000919.mm.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Inventoried Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Ucp1

Panther Classification:

Molecular Function -

transporter secondary carrier transporter

Gene Ontology Categories:

Function(s) Process(es)

regulation of transcription from RNA polymerase II promoter
transport
mitochondrial transport
proton transport
transmembrane transport
protein binding
oxidative phosphorylation uncoupler activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0800006790.mm.1 NM_009463.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0800006790.mm.1 NM_009463.3 Refseq
Affymetrix Mouse Clariom S Array PSR0800006790.mm.1 TC0800000919.mm.1 MTA TC

Back To Top
 

Citations (1)

Fibroblast growth factor 21 corrects obesity in mice.
Authors
Coskun T, Bina HA, Schneider MA, Dunbar JD, Hu CC, Chen Y, Moller DE, Kharitonenkov A
Journal
Endocrinology - Page No 6018 (Mon Dec 01 00:00:00 EST 2008)
Volume (Year)
149 (2008)
Product Usage
-

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-zpsr4:80/100.66.78.116:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0