This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01245336_g1
See other GPT2 GE Assays ›
Gene Symbol:
Gpt2
Gene Name:
glutamic pyruvate transaminase (alanine aminotransferase) 2
Gene Aliases:
4631422C05Rik, ALT2, AU014768, AU041193, C87201
Chromosome Location:
Chr.8: 85492474 - 85527560 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01245336_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Gpt2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_173866.3 NP_776291.1 8 - 9 1135 69
GenBank mRNA AK031958.1 - 8 - 9 1327 69
AK033424.1 - 8 - 9 1136 69
AK075894.1 - 8 - 9 1154 69
AK076250.1 - 8 - 9 1153 69
AK082030.1 - 8 - 9 1178 69
BC034219.1 - 8 - 9 1167 5065322 69

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Gpt2

Panther Classification:

Molecular Function -

transaminase

Gene Ontology Categories:

Function(s) Process(es)

2-oxoglutarate metabolic process
biosynthetic process
L-alanine metabolic process
catalytic activity
L-alanine:2-oxoglutarate aminotransferase activity
transaminase activity
transferase activity
pyridoxal phosphate binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-mvcqg:80/100.66.75.199:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0