This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01245525_g1
See other ELP5 GE Assays ›
Gene Symbol:
Elp5
Gene Name:
elongator acetyltransferase complex subunit 5
Gene Aliases:
Derp6, Rai12
Chromosome Location:
Chr.11: 69968224 - 69981294 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01245525_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Elp5
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001253700.1 NP_001240629.1 2 - 3 173 75
NM_018740.2 NP_061210.2 3 - 4 434 75
GenBank mRNA AA646690.1 - 2 - 3 89 75
AF004107.1 - 2 - 3 75
AK011130.1 - 1 - 2 246 75
AK152582.1 - 3 - 4 438 75
BC003752.1 - 2 - 3 285 3489878 75
BY292587.1 - 1 - 2 236 75

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the NM_001253700.1 transcripts.
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Elp5

Gene Ontology Categories:

Function(s)

transcription, DNA-templated
regulation of transcription, DNA-templated
positive regulation of cell migration

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-g27zl:80/100.66.78.232:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0