This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01245544_m1
See other EIF5A GE Assays ›
Gene Symbol:
Eif5a
Gene Name:
eukaryotic translation initiation factor 5A
Gene Aliases:
AA410058, D19Wsu54e, Eif4d, Eif5a1, eIF-4D, eIF-5A, eIF-5A-1, eIF-5A1
Chromosome Location:
Chr.11: 69916712 - 69921958 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01245544_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Eif5a
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001166594.1 NP_001160066.1 1 - 2 149 87
GenBank mRNA AY129328.1 - 1 - 2 142 87
CX218195.1 - 1 - 2 149 87

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the AY129328.1, NM_001166594.1 transcripts.

Set Membership:

Amplicon length between 86 and 100 Mus musculus Probe spans exons

Panther Classification:

Molecular Function -

translation initiation factor

Gene Ontology Categories:

Function(s) Process(es)

mRNA export from nucleus
translation
translational elongation
translational frameshifting
protein export from nucleus
transport
apoptotic process
positive regulation of cell proliferation
protein transport
positive regulation of apoptotic process
negative regulation of apoptotic process
positive regulation of translational elongation
positive regulation of translational termination
mRNA transport
positive regulation of muscle cell differentiation
RNA binding
translation elongation factor activity
U6 snRNA binding
ribosome binding
poly(A) RNA binding
protein N-terminus binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1100028293.mm.1 NM_001166589.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100028293.mm.1 NM_001166589.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100028293.mm.1 NM_001166590.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100028293.mm.1 NM_001166590.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100028293.mm.1 NM_001166591.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100028293.mm.1 NM_001166591.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100028293.mm.1 NM_001166592.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100028293.mm.1 NM_001166592.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100028293.mm.1 NM_001166593.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100028293.mm.1 NM_001166593.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100028293.mm.1 NM_001166594.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100028293.mm.1 NM_001166594.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100028293.mm.1 NM_001166595.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100028293.mm.1 NM_001166595.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100028293.mm.1 NM_001166596.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100028293.mm.1 NM_001166596.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100028293.mm.1 NM_181582.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100028293.mm.1 NM_181582.4 Refseq
Affymetrix Mouse Clariom S Array PSR1100028293.mm.1 TC1100003076.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1100028298.mm.1 NM_001166592.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100028298.mm.1 NM_001166592.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100028298.mm.1 NM_001166594.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100028298.mm.1 NM_001166594.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100028298.mm.1 TC1100003076.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-8rxhs:80/100.66.78.116:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0