This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01245704_m1
See other RNF167 GE Assays ›
Gene Symbol:
Rnf167
Gene Name:
ring finger protein 167
Gene Aliases:
0610010G05Rik, 5730408C10Rik, AU019305, AV328608
Chromosome Location:
Chr.11: 70647589 - 70651421 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01245704_m1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Rnf167
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AK017523.1 - 3 - 4 932 73

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length between 71 and 85 Mus musculus Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-hwq2n:80/100.66.77.213:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0