This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01245830_m1
See other SPNS3 GE Assays ›
Gene Symbol:
Spns3
Gene Name:
spinster homolog 3
Gene Aliases:
9830002I17Rik
Chromosome Location:
Chr.11: 72498153 - 72550508 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01245830_m1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Spns3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_029932.3 NP_084208.1 5 - 6 674 66
GenBank mRNA AK020683.1 - 5 - 6 675 66
AK041376.1 - 4 - 5 664 66
AK156479.1 - 5 - 6 938 66
AK156781.1 - 5 - 6 666 66
BC150778.1 - 5 - 6 657 9087689 66

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1100003147.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Transporter Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Spns3

Panther Classification:

Molecular Function -

transporter

Gene Ontology Categories:

Function(s) Process(es)

sphingosine-1-phosphate signaling pathway
transport
lipid transport
locomotion
transmembrane transport
molecular_function
sphingolipid transporter activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1100029210.mm.1 NM_029932.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100029210.mm.1 NM_029932.3 Refseq
Affymetrix Mouse Clariom S Array PSR1100029210.mm.1 TC1100003147.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-rpjph:80/100.66.75.107:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0