This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01246044_g1
See other CTNS GE Assays ›
Gene Symbol:
Ctns
Gene Name:
cystinosis, nephropathic
Gene Aliases:
AI195360, AW049661
Chromosome Location:
Chr.11: 73183133 - 73199029 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01246044_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Ctns
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_031251.4 NP_112541.1 5 - 6 451 115
GenBank mRNA AJ250670.1 - 4 - 5 349 115
AK054467.1 - 6 - 7 629 115
AK078511.1 - 5 - 6 463 115
AK079295.1 - 5 - 6 452 115
AK137143.1 - 5 - 6 494 115
AK170790.1 - 5 - 6 463 115
AK170828.1 - 5 - 6 474 115
BC005479.1 - 5 - 6 438 3484952 115
BC006033.1 - 5 - 6 438 115

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Transporter Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Ctns

Panther Classification:

Molecular Function -

amino acid transporter transporter secondary carrier transporter

Gene Ontology Categories:

Function(s) Process(es)

lens development in camera-type eye
glutathione metabolic process
transport
brain development
long-term memory
grooming behavior
adult walking behavior
visual learning
L-cystine transport
ATP metabolic process
cognition
L-alpha-amino acid transmembrane transport
L-cystine transmembrane transporter activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1100029275.mm.1 NM_031251.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100029275.mm.1 NM_031251.4 Refseq
Affymetrix Mouse Clariom S Array PSR1100029275.mm.1 TC1100003159.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-8rxhs:80/100.66.78.116:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0