This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01246078_g1
See other HDAC5 GE Assays ›
Gene Symbol:
Hdac5
Gene Name:
histone deacetylase 5
Gene Aliases:
AI426555, Hdac4, mHDA1, mKIAA0600
Chromosome Location:
Chr.11: 102195747 - 102230186 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01246078_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Hdac5
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001077696.1 NP_001071164.1 24 - 25 3431 95
NM_001284248.1 NP_001271177.1 26 - 27 3943 95
NM_001284249.1 NP_001271178.1 25 - 26 3540 95
NM_001284250.1 NP_001271179.1 24 - 25 3428 95
NM_010412.3 NP_034542.3 24 - 25 3428 95
GenBank mRNA AF006602.2 - 23 - 24 3141 95
AF207748.1 - 23 - 24 3060 95
AK122325.2 - 19 - 20 2470 95
AK139202.1 - 25 - 26 3515 95
AK146483.1 - 24 - 25 3367 95
AK155335.1 - 24 - 25 3402 95
AK156008.1 - 16 - 17 4812 95
AK157159.1 - 16 - 17 4812 95
AK161956.1 - 4 - 5 560 95
BC023309.1 - 15 - 16 1981 95
BC049926.1 - 24 - 25 3362 95
BC058554.1 - 26 - 27 3895 95
BC060609.1 - 23 - 24 3222 5720438 95

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Hdac5

Panther Classification:

Molecular Function -

histone modifying enzyme

Gene Ontology Categories:

Function(s) Process(es)

negative regulation of transcription from RNA polymerase II promoter
osteoblast development
transcription, DNA-templated
regulation of transcription, DNA-templated
transcription from RNA polymerase II promoter
protein deacetylation
inflammatory response
nervous system development
heart development
negative regulation of gene expression
regulation of myotube differentiation
negative regulation of myotube differentiation
chromatin modification
histone deacetylation
B cell differentiation
multicellular organismal response to stress
regulation of gene expression, epigenetic
B cell activation
response to cocaine
response to drug
regulation of protein binding
negative regulation of osteoblast differentiation
negative regulation of striated muscle tissue development
negative regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
regulation of skeletal muscle fiber development
positive regulation of sequence-specific DNA binding transcription factor activity
regulation of striated muscle cell differentiation
renal tubule morphogenesis
cellular response to fluid shear stress
negative regulation of cell migration involved in sprouting angiogenesis
positive regulation of neural precursor cell proliferation
positive regulation of stem cell proliferation
RNA polymerase III transcription factor binding
core promoter sequence-specific DNA binding
core promoter binding
transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
RNA polymerase II transcription factor binding
chromatin binding
transcription corepressor activity
histone deacetylase activity
protein kinase C binding
protein binding
transcription factor binding
hydrolase activity
protein kinase binding
histone deacetylase activity (H3-K14 specific)
NAD-dependent histone deacetylase activity (H3-K14 specific)
histone deacetylase activity (H3-K9 specific)
protein deacetylase activity
histone deacetylase activity (H4-K16 specific)
histone deacetylase binding
transcription regulatory region DNA binding
metal ion binding
NAD-dependent histone deacetylase activity (H3-K9 specific)
NAD-dependent histone deacetylase activity (H4-K16 specific)
repressing transcription factor binding
NAD-dependent histone deacetylase activity (H3-K18 specific)

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-mnfm6:80/100.66.78.37:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline