Assay Details
| Gene Symbol: Prkca | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011101.3 | NP_035231.2 | 6 - 7 | 903 | 71 | |
| GenBank mRNA | AK147084.1 | - | 6 - 7 | 904 | 71 | |
| BC096493.1 | - | 6 - 7 | 697 | 30742571 | 71 | |
| X52684.1 | - | 7 - 8 | 844 | 71 | ||
| X52685.1 | - | 7 - 8 | 844 | 71 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Prkca
Genomic Map