This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01246204_g1
See other NUDT4 GE Assays ›
Gene Symbol:
Nudt4
Gene Name:
nudix (nucleoside diphosphate linked moiety X)-type motif 4
Gene Aliases:
4933436C10Rik, DIPP2, DIPP2alpha, DIPP2beta, HDCMB47P
Chromosome Location:
Chr.10: 95547007 - 95564167 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01246204_g1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Nudt4
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_027722.4 NP_081998.3 2 - 3 619 61
GenBank mRNA AK017075.1 - 2 - 3 516 61

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Nudt4

Panther Classification:

Molecular Function -

phosphatase

Gene Ontology Categories:

Process(es)

RNA binding
thiamine-pyrophosphatase activity
8-oxo-7,8-dihydroguanosine triphosphate pyrophosphatase activity
diphosphoinositol-polyphosphate diphosphatase activity
UDP-2,3-diacylglucosamine hydrolase activity
bis(5'-nucleosyl)-tetraphosphatase activity
dATP pyrophosphohydrolase activity
pyrophosphatase activity
hydrolase activity
dihydroneopterin monophosphate phosphatase activity
dihydroneopterin triphosphate pyrophosphohydrolase activity
snoRNA binding
dITP diphosphatase activity
dTTP diphosphatase activity
XTP diphosphatase activity
ATP-dependent 5'-3' DNA helicase activity
phosphocholine hydrolase activity
metal ion binding
m7G(5')pppN diphosphatase activity
inositol diphosphate tetrakisphosphate diphosphatase activity
inositol bisdiphosphate tetrakisphosphate diphosphatase activity
inositol diphosphate pentakisphosphate diphosphatase activity
inositol-1-diphosphate-2,3,4,5,6-pentakisphosphate diphosphatase activity
inositol-3-diphosphate-1,2,4,5,6-pentakisphosphate diphosphatase activity
inositol-5-diphosphate-1,2,3,4,6-pentakisphosphate diphosphatase activity
inositol-1,5-bisdiphosphate-2,3,4,6-tetrakisphosphate 1-diphosphatase activity
inositol-1,5-bisdiphosphate-2,3,4,6-tetrakisphosphate 5-diphosphatase activity
inositol-3,5-bisdiphosphate-2,3,4,6-tetrakisphosphate 5-diphosphatase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-nbrrd:80/100.66.77.69:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0