This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01246215_m1
See other LTA4H GE Assays ›
Gene Symbol:
Lta4h
Gene Name:
leukotriene A4 hydrolase
Gene Aliases:
-
Chromosome Location:
Chr.10: 93453396 - 93484898 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01246215_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Lta4h
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_008517.2 NP_032543.2 11 - 12 1165 98
GenBank mRNA AK134931.1 - 11 - 12 1151 98
AK214707.1 - 2 - 3 120 98
AK216689.1 - 2 - 3 120 98
BC021417.1 - 11 - 12 1159 5004113 98
M63848.1 - 1131 98

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1000001131.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Protease Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Lta4h

Gene Ontology Categories:

Function(s) Process(es)

proteolysis
leukotriene metabolic process
leukotriene biosynthetic process
peptide catabolic process
aminopeptidase activity
epoxide hydrolase activity
leukotriene-A4 hydrolase activity
protein binding
peptidase activity
metallopeptidase activity
zinc ion binding
hydrolase activity
poly(A) RNA binding
metal ion binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1000008539.mm.1 NM_008517.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1000008539.mm.1 NM_008517.2 Refseq
Affymetrix Mouse Clariom S Array PSR1000008539.mm.1 TC1000001131.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-jmd5k:80/100.66.78.41:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0