This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01246489_g1
See other SGSM2 GE Assays ›
Gene Symbol:
Sgsm2
Gene Name:
small G protein signaling modulator 2
Gene Aliases:
D630003G22Rik, R74628, Rutbc1, mKIAA0397
Chromosome Location:
Chr.11: 74849262 - 74897093 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01246489_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Sgsm2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_197943.2 NP_922934.2 5 - 6 725 63
GenBank mRNA AK122273.1 - 5 - 6 647 63
BC058414.1 - 5 - 6 716 6822416 63
BC060163.1 - 5 - 6 716 6822416 63

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1100003204.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Sgsm2

Panther Classification:

Molecular Function -

GTPase-activating protein

Gene Ontology Categories:

Function(s) Process(es)

late endosome to Golgi transport
positive regulation of GTPase activity
GTPase activator activity
Rab GTPase binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1100029511.mm.1 NM_197943.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100029511.mm.1 NM_197943.2 Refseq
Affymetrix Mouse Clariom S Array PSR1100029511.mm.1 TC1100003204.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-mt7zw:80/100.66.79.31:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0