This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01246687_g1
See other ROMO1 GE Assays ›
Gene Symbol:
Romo1
Gene Name:
reactive oxygen species modulator 1
Gene Aliases:
2010100O12Rik, AI853864
Chromosome Location:
Chr.2: 156144153 - 156145794 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01246687_g1
Best Coverage
Size
Availability Inventoried
Catalog # 4331182
Price 284.00
Your Price
Online offer: 266.65
266.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Romo1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001164216.1 NP_001157688.1 2 - 3 264 85
NM_001164217.2 NP_001157689.1 2 - 3 278 85
NM_025946.6 NP_080222.1 1 - 2 362 85
GenBank mRNA AA058111.1 - 2 - 3 264 85
AK003488.1 - 2 - 3 173 85
AK008300.1 - 2 - 3 199 85
AY028425.1 - 1 - 2 257 85
BC028749.1 - 2 - 3 159 1196408 85
CJ221444.1 - 2 - 3 226 85
EL963857.1 - 1 - 2 372 85

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Romo1

Gene Ontology Categories:

Function(s) Process(es)

replicative cell aging
positive regulation of cell proliferation
cellular response to reactive oxygen species
defense response to bacterium
positive regulation of reactive oxygen species metabolic process
molecular_function

MicroArray Probes:

Agilent Affymetrix ABI Illumina

Array Name Probe IDs Targets Target Type
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21769698 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21769695 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21769697 - -
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1980341 NM_025946.5 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1980341 AK003488.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1980341 AK008300.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1980341 AY028425.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1980341 BC028749.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1989276 NM_025946.5 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1989276 AA058111.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1989276 AK003488.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1989276 AK008300.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1989276 AY028425.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2114467 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 853648 - -
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1989276 NM_025946.5 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1989276 AA058111.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1989276 AK003488.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1989276 AK008300.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1989276 AY028425.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2114467 - -
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1980341 NM_025946.5 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1980341 AK003488.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1980341 AK008300.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1980341 AY028425.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1980341 BC028749.1 mouse_transcriptome
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21769695 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21769703 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21769698 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21769704 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21769699 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21769700 - -
Whole Mouse Genome G4122A A_51_P199148 NM_025946.5 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P199148 AA058111.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P199148 AK003488.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P199148 AK008300.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P199148 AY028425.1 mouse_transcriptome
Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P114447 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01887024 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01887020 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01887005 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01887013 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21769695 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21769697 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21769698 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21769710 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21769695 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21769694 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21769700 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21769699 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21769714 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21769712 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21769701 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21769708 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21769699 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P01887020 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21769703 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21769697 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21769708 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21769704 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21769710 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21769714 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21769712 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P01293325 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21769695 - -
Array Name Probe IDs Targets Target Type
Affymetrix Mouse Gene 1.0 ST Array 10477812 - -
Affymetrix Mouse Gene 1.0 ST Array 10477809 - -
Affymetrix Mouse Gene 1.0 ST Array 10495253 - -
Affymetrix Mouse Gene 1.0 ST Array 10477811 - -
Affymetrix Mouse Gene 1.0 ST Array 10477810 - -
Affymetrix HT MG-430 PM Array Plate 1420642_PM_a_at - -
Affymetrix Mouse Gene 1.1 ST Array 10495252 - -
Affymetrix Mouse Gene 1.1 ST Array 10477808 - -
Mouse Genome 430 2.0 Array 1420642_PM_a_at - -
Mouse Genome 430 2.0 Array 1420642_a_at - -
Affymetrix GeneChip Mouse Genome 430 2.0 Array 67067_PM_at - -
Affymetrix GeneChip Mouse Genome 430 2.0 Array 67067_at - -
Affymetrix Mouse Clariom S Array PSR0200019158.mm.1 NM_001164216.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200019158.mm.1 NM_001164216.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200019158.mm.1 NM_001164217.2 Refseq
Affymetrix Mouse Clariom S Array PSR0200019158.mm.1 NM_025946.6 Refseq
Affymetrix Mouse Clariom S Array PSR0200019158.mm.1 TC0200002384.mm.1 MTA TC
Mouse Expression Set 430 1420642_a_at - -
Mouse Expression Set 430 1420642_PM_a_at - -
Murine Genome U74v2 Set 96687_f_at - -
Murine Genome U74v2 Set 96687_PM_f_at - -
Mouse Genome 430A 2.0 Array 1420642_PM_a_at - -
Mouse Genome 430A 2.0 Array 1420642_a_at - -
Affymetrix HT Mouse Genome 430A Array 1420642_PM_a_at - -
Affymetrix HT Mouse Genome 430A Array 1420642_a_at - -
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 897170 - -
Array Name Probe IDs Targets Target Type
MouseWG-6 v2 Expression BeadChip 1940095 - -
Illumina mouse-6 v1.0 expression beadchip 1940095 - -
Illumina mouse-6 v1.0 expression beadchip 770537 BC145847.1 genbank_mrna_mouse
Illumina mouse-6 v1.0 expression beadchip 770537 NM_146683.1 refseq_mrna_mouse_NM
Illumina mouseRef-8 v1.0 expression beadchip 0000770537 - -
Illumina mouseRef-8 v1.0 expression beadchip 0001940095 - -
Illumina mouseRef-8 v1.1 expression beadchip ILMN_2670427 - -
Illumina mouseRef-8 v1.1 expression beadchip ILMN_1228094 - -
Illumina mouse-6 v1.1 expression beadchip ILMN_1228094 - -
Illumina mouse-6 v1.1 expression beadchip ILMN_2670427 - -
mouse-6 v1.1 (Illumina) scl0067067.1_41-S - -
mouse-6 v1.1 (Illumina) scl067067.1_49-S - -
Illumina Sentrix mouse-6.v1 bead array 1940095 - -
Illumina Sentrix mouse-6.v1 bead array 770537 BC145847.1 genbank_mrna_mouse
Illumina Sentrix mouse-6.v1 bead array 770537 NM_146683.1 refseq_mrna_mouse_NM
Illumina Sentrix MouseRef-8 Expression BeadChip scl067067.1_49-S - -
Illumina Sentrix MouseRef-8 Expression BeadChip scl0067067.1_41-S - -
Illumina Sentrix Mouse-6 Expression BeadChip (2) 1940095 - -
Illumina Sentrix Mouse-6 Expression BeadChip (2) 770537 BC145847.1 genbank_mrna_mouse
Illumina Sentrix Mouse-6 Expression BeadChip (2) 770537 NM_146683.1 refseq_mrna_mouse_NM
MouseRef-8 v2 Expression BeadChip ILMN_1228094 - -
MouseRef-8 v2 Expression BeadChip ILMN_2670427 - -
Illumina MouseWG-6 v2.0 expression beadchip ILMN_1228094 - -
Illumina MouseWG-6 v2.0 expression beadchip ILMN_2670427 - -
Illumina Sentrix Mouse-6 Expression BeadChip (3) scl067067.1_49-S - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-mt7zw:80/100.66.79.31:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0