This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01246827_g1
See other ACOX1 GE Assays ›
Gene Symbol:
Acox1
Gene Name:
acyl-Coenzyme A oxidase 1, palmitoyl
Gene Aliases:
AOX, Acox, D130055E20Rik, Paox
Chromosome Location:
Chr.11: 116171883 - 116199045 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01246827_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Acox1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001271898.1 NP_001258827.1 12 - 13 2063 68
NM_015729.3 NP_056544.2 12 - 13 2063 68
GenBank mRNA AB034914.1 - 12 - 13 1752 68
AF006688.1 - 12 - 13 1802 68
AK034760.1 - 8 - 9 1368 68
AK040566.1 - 12 - 13 1766 68
AK128941.1 - 12 - 13 1765 68
AK170217.1 - 8 - 9 1133 68
BC054727.1 - 12 - 13 1731 68
BC056448.1 - 12 - 13 2063 5704873 68

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1100004107.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Acox1

Panther Classification:

Molecular Function -

oxidoreductase

Gene Ontology Categories:

Function(s) Process(es)

very long-chain fatty acid metabolic process
generation of precursor metabolites and energy
lipid metabolic process
fatty acid metabolic process
fatty acid beta-oxidation
prostaglandin metabolic process
spermatogenesis
metabolic process
peroxisome fission
fatty acid oxidation
fatty acid beta-oxidation using acyl-CoA oxidase
lipid homeostasis
oxidation-reduction process
positive regulation of cholesterol homeostasis
acyl-CoA dehydrogenase activity
acyl-CoA oxidase activity
receptor binding
fatty acid binding
palmitoyl-CoA oxidase activity
oxidoreductase activity
oxidoreductase activity, acting on the CH-CH group of donors
PDZ domain binding
protein N-terminus binding
flavin adenine dinucleotide binding
FAD binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-mnfm6:80/100.66.78.37:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline