This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01246988_g1
See other PITPNC1 GE Assays ›
Gene Symbol:
Pitpnc1
Gene Name:
phosphatidylinositol transfer protein, cytoplasmic 1
Gene Aliases:
1110020B03Rik, 5830436L09Rik, AI662802, AI851387, C330017I21Rik, Dnr411, RDGBB, RDGBB1, rdgB-beta
Chromosome Location:
Chr.11: 107207892 - 107470892 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01246988_g1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Pitpnc1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_145823.2 NP_665822.1 7 - 8 1397 64
GenBank mRNA AB077281.1 - 7 - 8 1378 64
AB077282.1 - 7 - 8 675 64
AK003844.1 - 6 - 7 645 64
AK171340.1 - 7 - 8 1424 64
AK171918.1 - 7 - 8 1394 64
BC082333.1 - 7 - 8 1229 64
BC108351.1 - 7 - 8 1257 6312193 64

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Transporter Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Pitpnc1

Panther Classification:

Molecular Function -

transporter

Gene Ontology Categories:

Function(s) Process(es)

transport
lipid transport
phospholipid transport
phosphatidylinositol metabolic process
lipid binding
phosphatidylinositol transporter activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1100037139.mm.1 NM_145823.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100037139.mm.1 NM_145823.2 Refseq
Affymetrix Mouse Clariom S Array PSR1100037139.mm.1 TC1100003948.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1100037140.mm.1 NM_145823.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100037140.mm.1 NM_145823.2 Refseq
Affymetrix Mouse Clariom S Array PSR1100037140.mm.1 TC1100003948.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-rpjph:80/100.66.75.107:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0