This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01247092_g1
See other CCT2 GE Assays ›
Gene Symbol:
Cct2
Gene Name:
chaperonin containing Tcp1, subunit 2 (beta)
Gene Aliases:
Cctb
Chromosome Location:
Chr.10: 117050997 - 117063814 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01247092_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Cct2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_007636.2 NP_031662.2 5 - 6 441 93
GenBank mRNA AB041570.1 - 5 - 6 413 93
AK012295.1 - 5 - 6 397 93
AK075581.1 - 5 - 6 414 93
AK088145.1 - 5 - 6 403 93
AK145406.1 - 5 - 6 442 93
AK166891.1 - 5 - 6 414 93
AK179930.1 - 3 - 4 215 93
BC007470.1 - 5 - 6 405 3601569 93
BC026918.1 - 5 - 6 407 4500896 93
Z31553.1 - 5 - 6 392 93

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1000002920.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Cct2

Panther Classification:

Molecular Function -

chaperonin

Gene Ontology Categories:

Function(s) Process(es)

protein folding
binding of sperm to zona pellucida
chaperone-mediated protein complex assembly
toxin transport
nucleotide binding
ATP binding
ubiquitin protein ligase binding
unfolded protein binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1000022079.mm.1 NM_007636.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1000022079.mm.1 NM_007636.2 Refseq
Affymetrix Mouse Clariom S Array PSR1000022079.mm.1 TC1000002920.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1000022080.mm.1 NM_007636.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1000022080.mm.1 NM_007636.2 Refseq
Affymetrix Mouse Clariom S Array PSR1000022080.mm.1 TC1000002920.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-w8bxd:80/100.66.79.51:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0