This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01247310_m1
See other CDH13 GE Assays ›
Gene Symbol:
Cdh13
Gene Name:
cadherin 13
Gene Aliases:
4932416G01Rik, Cdht, T-cadherin, Tcad
Chromosome Location:
Chr.8: 118032249 - 119324929 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01247310_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Cdh13
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_019707.4 NP_062681.2 1 - 2 147 90
GenBank mRNA AB022100.1 - 1 - 2 156 90
AK029195.1 - 1 - 2 106 90
AK032409.1 - 1 - 2 176 90
AK034995.1 - 1 - 2 127 90
AK039438.1 - 1 - 2 169 90
AK045201.1 - 1 - 2 148 90
AK048724.1 - 1 - 2 147 90
AK085939.1 - 1 - 2 107 90
AK134649.1 - 1 - 2 106 90
BC021628.1 - 1 - 2 161 3987627 90

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the AB022100.1, AK029195.1, AK034995.1, AK039438.1, AK048724.1, AK134649.1, BC021628.1, NM_019707.4 transcripts.

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus 1700 Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Cdh13

Panther Classification:

Molecular Function -

cadherin

Gene Ontology Categories:

Function(s) Process(es)

regulation of cell growth
positive regulation of endothelial cell proliferation
positive regulation of cell-matrix adhesion
sprouting angiogenesis
cell adhesion
homophilic cell adhesion via plasma membrane adhesion molecules
negative regulation of cell adhesion
Rho protein signal transduction
negative regulation of cell proliferation
calcium-dependent cell-cell adhesion via plasma membrane cell adhesion molecules
Rac protein signal transduction
lamellipodium assembly
regulation of endocytosis
positive regulation of cell migration
regulation of epidermal growth factor receptor signaling pathway
endothelial cell migration
keratinocyte proliferation
positive regulation of transcription from RNA polymerase II promoter
positive regulation of smooth muscle cell proliferation
positive regulation of calcium-mediated signaling
positive regulation of positive chemotaxis
localization within membrane
low-density lipoprotein particle mediated signaling
calcium ion binding
protein binding
low-density lipoprotein particle binding
protein homodimerization activity
cadherin binding
metal ion binding
adiponectin binding
lipoprotein particle binding

MicroArray Probes:

ABI

Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 630644 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-wt24k:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline