This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01247314_m1
See other CDH13 GE Assays ›
Gene Symbol:
Cdh13
Gene Name:
cadherin 13
Gene Aliases:
4932416G01Rik, Cdht, T-cadherin, Tcad
Chromosome Location:
Chr.8: 118032249 - 119324929 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01247314_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Cdh13
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_019707.4 NP_062681.2 5 - 6 739 75
GenBank mRNA AB022100.1 - 5 - 6 748 75
AK029195.1 - 5 - 6 698 75
AK034995.1 - 5 - 6 719 75
AK039438.1 - 5 - 6 761 75
AK048724.1 - 5 - 6 739 75
AK134649.1 - 5 - 6 698 75
BC021628.1 - 5 - 6 753 3987627 75

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Cdh13

Panther Classification:

Molecular Function -

cadherin

Gene Ontology Categories:

Function(s) Process(es)

regulation of cell growth
positive regulation of endothelial cell proliferation
positive regulation of cell-matrix adhesion
sprouting angiogenesis
cell adhesion
homophilic cell adhesion via plasma membrane adhesion molecules
negative regulation of cell adhesion
Rho protein signal transduction
negative regulation of cell proliferation
calcium-dependent cell-cell adhesion via plasma membrane cell adhesion molecules
Rac protein signal transduction
lamellipodium assembly
regulation of endocytosis
positive regulation of cell migration
regulation of epidermal growth factor receptor signaling pathway
endothelial cell migration
keratinocyte proliferation
positive regulation of transcription from RNA polymerase II promoter
positive regulation of smooth muscle cell proliferation
positive regulation of calcium-mediated signaling
positive regulation of positive chemotaxis
localization within membrane
low-density lipoprotein particle mediated signaling
calcium ion binding
protein binding
low-density lipoprotein particle binding
protein homodimerization activity
cadherin binding
metal ion binding
adiponectin binding
lipoprotein particle binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-97tfg:80/100.66.76.242:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline