This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01247356_m1
See other CDH1 GE Assays ›
Gene Symbol:
Cdh1
Gene Name:
cadherin 1
Gene Aliases:
AA960649, E-cad, Ecad, L-CAM, UVO, Um
Chromosome Location:
Chr.8: 106603368 - 106670246 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01247356_m1
Size
Availability Inventoried
Catalog # 4331182
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Cdh1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_009864.2 NP_033994.1 8 - 9 1268 87
GenBank mRNA AK076369.1 - 8 - 9 1270 87
AK166267.1 - 8 - 9 1269 87

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mus musculus Inventoried Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Cdh1

Panther Classification:

Molecular Function -

cadherin

Gene Ontology Categories:

Function(s) Process(es)

in utero embryonic development
trophectodermal cell differentiation
epithelial cell morphogenesis
cell adhesion
homophilic cell adhesion via plasma membrane adhesion molecules
synapse assembly
sensory perception of sound
single organismal cell-cell adhesion
calcium-dependent cell-cell adhesion via plasma membrane cell adhesion molecules
protein metabolic process
negative regulation of cell-cell adhesion
regulation of protein localization
regulation of water loss via skin
adherens junction organization
positive regulation of transcription factor import into nucleus
regulation of cysteine-type endopeptidase activity involved in apoptotic process
positive regulation of transcription, DNA-templated
regulation of epithelial cell proliferation
negative regulation of epithelial cell proliferation
protein homooligomerization
intestinal epithelial cell development
salivary gland cavitation
regulation of branching involved in salivary gland morphogenesis
bicellular tight junction assembly
cellular response to amino acid stimulus
cellular response to lithium ion
cellular response to indole-3-methanol
protein localization to plasma membrane
establishment of protein localization to plasma membrane
negative regulation of canonical Wnt signaling pathway
cochlea development
regulation of protein localization to cell surface
regulation of neuron migration
glycoprotein binding
calcium ion binding
protein binding
beta-catenin binding
protein phosphatase binding
protein domain specific binding
ankyrin binding
GTPase activating protein binding
gamma-catenin binding
metal ion binding
cell adhesion molecule binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-ng27p:80/100.66.78.145:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline