This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01247382_m1
See other WWOX GE Assays ›
Gene Symbol:
Wwox
Gene Name:
WW domain-containing oxidoreductase
Gene Aliases:
5330426P09Rik, 9030416C10Rik, WOX1
Chromosome Location:
Chr.8: 114439630 - 115352712 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01247382_m1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Wwox
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_019573.3 NP_062519.2 3 - 4 350 82
GenBank mRNA AF187014.1 - 3 - 4 301 82
AK018507.1 - 3 - 4 280 82
AK019911.1 - 3 - 4 308 82
AK046903.1 - 3 - 4 300 82
AK078528.1 - 3 - 4 303 82
AK083096.1 - 3 - 4 282 82
BC014716.1 - 3 - 4 306 4241066 82
CJ058964.1 - 3 - 4 351 82

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0800001362.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Apoptosis Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Wwox

Panther Classification:

Molecular Function -

transcription cofactor

Gene Ontology Categories:

Function(s) Process(es)

osteoblast differentiation
apoptotic process
metabolic process
Wnt signaling pathway
negative regulation of Wnt signaling pathway
positive regulation of transcription from RNA polymerase II promoter
skeletal system morphogenesis
oxidation-reduction process
cellular response to transforming growth factor beta stimulus
intrinsic apoptotic signaling pathway by p53 class mediator
extrinsic apoptotic signaling pathway
positive regulation of extrinsic apoptotic signaling pathway
positive regulation of extrinsic apoptotic signaling pathway in absence of ligand
C-3 sterol dehydrogenase (C-4 sterol decarboxylase) activity
RNA polymerase II transcription coactivator activity
aldo-keto reductase (NADP) activity
isocitrate dehydrogenase activity
mevaldate reductase activity
protein binding
gluconate dehydrogenase activity
steroid dehydrogenase activity
oxidoreductase activity
epoxide dehydrogenase activity
5-exo-hydroxycamphor dehydrogenase activity
2-hydroxytetrahydrofuran dehydrogenase activity
acetoin dehydrogenase activity
enzyme binding
phenylcoumaran benzylic ether reductase activity
D-xylose:NADP reductase activity
L-arabinose:NADP reductase activity
D-arabinitol dehydrogenase, D-ribulose forming (NADP+) activity
steroid dehydrogenase activity, acting on the CH-OH group of donors, NAD or NADP as acceptor
steroid dehydrogenase activity, acting on the CH-CH group of donors
(R)-(-)-1,2,3,4-tetrahydronaphthol dehydrogenase activity
3-hydroxymenthone dehydrogenase activity
very long-chain-3-hydroxyacyl-CoA dehydrogenase activity
dihydrotestosterone 17-beta-dehydrogenase activity
(R)-2-hydroxyisocaproate dehydrogenase activity
L-arabinose 1-dehydrogenase (NADP+) activity
L-xylulose reductase (NAD+) activity
3-ketoglucose-reductase activity
(R)-2-hydroxyglutarate dehydrogenase activity
D-arabinitol dehydrogenase, D-xylulose forming (NADP+) activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0800011208.mm.1 NM_019573.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0800011208.mm.1 NM_019573.3 Refseq
Affymetrix Mouse Clariom S Array PSR0800011208.mm.1 TC0800001362.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0800011210.mm.1 NM_019573.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0800011210.mm.1 NM_019573.3 Refseq
Affymetrix Mouse Clariom S Array PSR0800011210.mm.1 TC0800001362.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0800011207.mm.1 NM_019573.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0800011207.mm.1 NM_019573.3 Refseq
Affymetrix Mouse Clariom S Array PSR0800011207.mm.1 TC0800001362.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0800011203.mm.1 NM_019573.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0800011203.mm.1 NM_019573.3 Refseq
Affymetrix Mouse Clariom S Array PSR0800011203.mm.1 TC0800001362.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0800011206.mm.1 NM_019573.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0800011206.mm.1 NM_019573.3 Refseq
Affymetrix Mouse Clariom S Array PSR0800011206.mm.1 TC0800001362.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0800011209.mm.1 NM_019573.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0800011209.mm.1 NM_019573.3 Refseq
Affymetrix Mouse Clariom S Array PSR0800011209.mm.1 TC0800001362.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-fcf5f9d6d-dcxfx:80/100.66.77.77:80.
git-commit: 856de8fa7eff2d55603d92afdf035972e45c3e8a
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.51.0-2026.05.21-1.0