This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01247569_m1
See other MON1B GE Assays ›
Gene Symbol:
Mon1b
Gene Name:
MON1 homolog b (yeast)
Gene Aliases:
5031407H10, 5033413H12Rik
Chromosome Location:
Chr.8: 113635586 - 113645192 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01247569_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Mon1b
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001048143.2 NP_001041608.1 5 - 6 1592 119
NM_173015.3 NP_766603.1 4 - 5 1782 119
GenBank mRNA AK030288.1 - 4 - 5 1748 119
AK157390.1 - 5 - 6 1595 119
AK173047.1 - 4 - 5 1454 119
BC062639.1 - 4 - 5 1458 119
BQ831297.1 - 2 - 3 195 119

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0800001357.mm.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Mus musculus 1700 Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Mon1b

Gene Ontology Categories:

Function(s) Process(es)

biological_process
vesicle-mediated transport
molecular_function

MicroArray Probes:

Agilent Affymetrix ABI Illumina

Array Name Probe IDs Targets Target Type
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26179660 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26179657 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26179658 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26179661 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26179659 - -
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2076471 AK173047.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2076471 BC062639.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2076471 NM_001048143.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2076471 NM_173015.2 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2076471 AK017167.1 mouse_transcriptome
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 32300 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 645635 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 657280 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 649097 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 634339 - -
Agilent SurePrint G3 Mouse CGH Microarray 4x180K 76487 - -
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2076471 AK173047.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2076471 BC062639.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2076471 NM_001048143.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2076471 NM_173015.2 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2076471 AK017167.1 mouse_transcriptome
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26179676 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26179650 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26179642 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26179658 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26179674 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26179678 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26179669 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26179656 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26179664 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26179661 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26179634 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26179681 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26179672 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26179657 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26179626 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26179660 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26179631 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26179636 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26179648 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26179649 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26179640 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26179652 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26179654 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26179638 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26179647 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26179682 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26179644 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26179668 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26179677 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26179632 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26179671 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26179675 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26179627 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P06647277 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P01832134 - -
Whole Mouse Genome G4122A A_52_P143757 AK157390.1 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P143757 AK173047.1 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P143757 BC062639.1 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P143757 BQ831297.1 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P143757 NM_001048143.1 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P56606 AK173047.1 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P56606 BC062639.1 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P56606 NM_001048143.1 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P56606 NM_173015.2 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P56606 AK017167.1 mouse_transcriptome
Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P102074 - -
Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P06647277 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07045112 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07045111 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07045123 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07045116 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26179661 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26179659 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26179660 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26179657 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26179658 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26179778 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26179769 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26179599 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26179709 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26179719 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26179618 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26179780 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26179647 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26179654 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26179602 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26179741 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26179617 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26179783 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26179607 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26179807 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26179722 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26179649 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26179801 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26179743 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26179658 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26179798 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26179784 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26179800 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26179767 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26179764 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26179640 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26179728 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26179660 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26179787 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26179773 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26179790 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26179771 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26179706 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26179750 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26179803 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26179623 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26179631 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26179736 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26179621 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26179774 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26179752 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26179782 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26179711 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26179738 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26179793 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26179627 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26179761 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26179714 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26179789 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26179777 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26179626 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26179768 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26179765 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26179708 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26179759 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26179612 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26179730 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26179634 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26179702 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26179717 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26179611 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26179729 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26179733 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26179636 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26179726 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26179652 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26179661 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26179754 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26179712 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26179757 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26179788 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26179747 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26179648 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26179760 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26179641 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26179744 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26179724 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26179795 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26179609 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26179701 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26179775 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26179698 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26179758 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26179656 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26179615 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26179805 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26179605 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26179626 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P07125536 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26179634 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26179621 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26179649 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P07208506 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26179660 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26179652 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P07287873 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26179627 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26179648 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26179640 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26179647 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26179662 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P07031251 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26179645 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26179658 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P07045123 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26179620 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P07125540 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26179613 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26179618 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26179639 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26179624 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26179656 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26179623 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26179636 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P06951523 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26179615 - -
Array Name Probe IDs Targets Target Type
Affymetrix Mouse Gene 1.0 ST Array 10575668 - -
Affymetrix Mouse Gene 1.0 ST Array 10575666 - -
Affymetrix Mouse Gene 1.0 ST Array 10575671 - -
Affymetrix Mouse Gene 1.0 ST Array 10575673 - -
Affymetrix Mouse Gene 1.0 ST Array 10575663 - -
Affymetrix Mouse Gene 1.0 ST Array 10575669 - -
Affymetrix Mouse Gene 1.0 ST Array 10575665 - -
Affymetrix Mouse Gene 1.0 ST Array 10575670 - -
Affymetrix Mouse Gene 1.0 ST Array 10575667 - -
Affymetrix Mouse Gene 1.0 ST Array 10575664 - -
Affymetrix HT MG-430 PM Array Plate 1432685_PM_at - -
Affymetrix HT MG-430 PM Array Plate 1428712_PM_at - -
Affymetrix HT MG-430 PM Array Plate 1428711_PM_at - -
Affymetrix Mouse Gene 1.1 ST Array 10575672 - -
Affymetrix Mouse Gene 1.1 ST Array 10575662 - -
Mouse Genome 430 2.0 Array 1428711_PM_at - -
Mouse Genome 430 2.0 Array 1428711_at - -
Mouse Genome 430 2.0 Array 1428712_at - -
Mouse Genome 430 2.0 Array 1428712_PM_at - -
Mouse Genome 430 2.0 Array 1432685_PM_at - -
Mouse Genome 430 2.0 Array 1432685_at - -
Affymetrix GeneChip Mouse Genome 430 2.0 Array 270096_PM_at - -
Affymetrix GeneChip Mouse Genome 430 2.0 Array 270096_at - -
Affymetrix Mouse Clariom S Array PSR0800011140.mm.1 NM_001048143.2 Refseq
Affymetrix Mouse Clariom S Array PSR0800011140.mm.1 NM_173015.3 Refseq
Affymetrix Mouse Clariom S Array PSR0800011140.mm.1 TC0800001357.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0800011136.mm.1 NM_001048143.2 Refseq
Affymetrix Mouse Clariom S Array PSR0800011136.mm.1 NM_173015.3 Refseq
Affymetrix Mouse Clariom S Array PSR0800011136.mm.1 TC0800001357.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0800011137.mm.1 NM_001048143.2 Refseq
Affymetrix Mouse Clariom S Array PSR0800011137.mm.1 NM_173015.3 Refseq
Affymetrix Mouse Clariom S Array PSR0800011137.mm.1 TC0800001357.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0800011139.mm.1 NM_001048143.2 Refseq
Affymetrix Mouse Clariom S Array PSR0800011139.mm.1 NM_173015.3 Refseq
Affymetrix Mouse Clariom S Array PSR0800011139.mm.1 TC0800001357.mm.1 MTA TC
Mouse Expression Set 430 1428711_PM_at - -
Mouse Expression Set 430 1428712_at - -
Mouse Expression Set 430 1428711_at - -
Mouse Expression Set 430 1428712_PM_at - -
Affymetrix HT Mouse Genome 430B Array 1428712_at - -
Affymetrix HT Mouse Genome 430B Array 1428712_PM_at - -
Affymetrix HT Mouse Genome 430B Array 1428711_PM_at - -
Affymetrix HT Mouse Genome 430B Array 1428711_at - -
Affymetrix HT Mouse Genome 430B Array 1432685_PM_at - -
Affymetrix HT Mouse Genome 430B Array 1432685_at - -
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 829355 - -
Array Name Probe IDs Targets Target Type
MouseWG-6 v2 Expression BeadChip 5890037 - -
MouseWG-6 v2 Expression BeadChip 102480270 - -
Illumina mouse-6 v1.0 expression beadchip 3520373 - -
Illumina mouse-6 v1.0 expression beadchip 5890037 - -
Illumina mouse-6 v1.0 expression beadchip 102480270 - -
Illumina mouseRef-8 v1.0 expression beadchip 0003520373 - -
Illumina mouseRef-8 v1.0 expression beadchip 0005890037 - -
Illumina mouseRef-8 v1.1 expression beadchip ILMN_2594558 - -
Illumina mouseRef-8 v1.1 expression beadchip ILMN_1219118 - -
Illumina mouse-6 v1.1 expression beadchip ILMN_1219118 - -
Illumina mouse-6 v1.1 expression beadchip ILMN_1219281 - -
Illumina mouse-6 v1.1 expression beadchip ILMN_2594558 - -
mouse-6 v1.1 (Illumina) scl00270096.1_196-S - -
mouse-6 v1.1 (Illumina) scl0270096.5_118-S - -
mouse-6 v1.1 (Illumina) scl33304.1.1_129-S - -
Illumina Sentrix mouse-6.v1 bead array 3520373 - -
Illumina Sentrix mouse-6.v1 bead array 102480270 - -
Illumina Sentrix mouse-6.v1 bead array 5890037 - -
Illumina Sentrix MouseRef-8 Expression BeadChip scl00270096.1_196-S - -
Illumina Sentrix MouseRef-8 Expression BeadChip scl0270096.5_118-S - -
Illumina Sentrix Mouse-6 Expression BeadChip (2) 5890037 - -
Illumina Sentrix Mouse-6 Expression BeadChip (2) 3520373 - -
Illumina Sentrix Mouse-6 Expression BeadChip (2) 102480270 - -
MouseRef-8 v2 Expression BeadChip ILMN_1219118 - -
MouseRef-8 v2 Expression BeadChip ILMN_3053203 - -
MouseRef-8 v2 Expression BeadChip ILMN_2594558 - -
Illumina MouseWG-6 v2.0 expression beadchip ILMN_1219118 - -
Illumina MouseWG-6 v2.0 expression beadchip ILMN_2594558 - -
Illumina MouseWG-6 v2.0 expression beadchip ILMN_3053203 - -
Illumina Sentrix Mouse-6 Expression BeadChip (3) scl00270096.1_196-S - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-gsg6q:80/100.66.76.182:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0