This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01247579_g1
See other ADGRG5 GE Assays ›
Gene Symbol:
Adgrg5
Gene Name:
adhesion G protein-coupled receptor G5
Gene Aliases:
Gm1109, Gpr114, PGR27
Chromosome Location:
Chr.8: 94921811 - 94943293 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01247579_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Adgrg5
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001033468.3 NP_001028640.2 11 - 12 1549 76
NM_001145972.1 NP_001139444.1 11 - 12 1557 76
GenBank mRNA AK028878.1 - 11 - 12 1550 76

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0800001125.mm.1

Set Membership:

Amplicon length between 71 and 85 G-protein coupled receptor 3' Most Detect Genomic DNA Mus musculus 1700 Probe spans exons

Panther Classification:

Molecular Function -

G-protein coupled receptor

Gene Ontology Categories:

Function(s) Process(es)

signal transduction
cell surface receptor signaling pathway
G-protein coupled receptor signaling pathway
signal transducer activity
transmembrane signaling receptor activity
G-protein coupled receptor activity

MicroArray Probes:

Agilent Affymetrix ABI Illumina

Array Name Probe IDs Targets Target Type
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2156638 BY547428.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2156638 AK156609.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2156638 AK028878.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2156638 AK156147.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2156638 NM_001033468.3 mouse_transcriptome
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 763291 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 624135 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 587842 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 186075 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 168 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 610898 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 358226 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 75077 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 334121 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 541780 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 333963 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 683794 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 766535 - -
Agilent SurePrint G3 Mouse CGH Microarray 4x180K 17732 - -
Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P165086 - -
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2156638 BY547428.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2156638 AK156609.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2156638 AK028878.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2156638 AK156147.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2156638 NM_001033468.3 mouse_transcriptome
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26080295 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26080282 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26080291 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26080290 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26080286 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26080283 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26080287 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26080294 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26080298 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26080285 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26080288 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P01802027 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P165086 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P06603960 - -
Whole Mouse Genome G4122A A_52_P200686 AK156609.1 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P200686 AY255569.1 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P200686 BC144851.1 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P200686 BC145803.1 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P200686 AK028878.1 mouse_transcriptome
Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P165086 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26080273 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26080311 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26080308 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26080217 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26080278 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26080349 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26080415 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26080414 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26080282 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26080270 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26080336 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26080252 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26080286 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26080339 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26080356 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26080224 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26080219 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26080231 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26080223 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26080259 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26080383 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26080216 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26080264 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26080294 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26080288 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26080245 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26080345 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26080326 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26080214 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26080391 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26080319 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26080422 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26080369 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26080429 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26080250 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26080358 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26080333 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26080235 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26080271 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26080242 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26080375 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26080279 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26080396 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26080277 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26080401 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26080275 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26080344 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26080363 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26080432 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26080254 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26080253 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26080437 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26080266 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26080412 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26080342 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26080328 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26080299 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26080227 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26080418 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26080351 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26080274 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26080389 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26080379 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26080324 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26080400 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26080419 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26080354 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26080330 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26080283 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26080291 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26080397 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26080322 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26080410 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26080427 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26080285 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26080222 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26080263 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26080337 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26080353 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26080387 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26080226 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26080367 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26080385 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26080211 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26080406 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26080281 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26080247 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26080261 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26080241 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26080381 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26080240 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26080309 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26080220 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26080314 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26080402 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26080233 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26080228 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26080377 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26080404 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26080287 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26080425 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26080434 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26080361 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26080359 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26080237 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P06769659 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P06874945 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26080285 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26080279 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P07124066 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26080410 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26080275 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26080400 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P06823179 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26080274 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P07124063 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P06769658 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P07304697 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26080305 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26080307 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P07124074 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26080300 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26080301 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P07124067 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26080401 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P07022788 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26080273 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26080281 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P07124059 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P07124070 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26080270 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26080291 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P07124078 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P06769657 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26080277 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26080298 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P06728986 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P06683288 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26080288 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26080412 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26080306 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26080278 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P06769660 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26080299 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P07150262 - -
Array Name Probe IDs Targets Target Type
Affymetrix Mouse Gene 1.0 ST Array 10574257 - -
Affymetrix Mouse Gene 1.0 ST Array 10574253 - -
Affymetrix Mouse Gene 1.0 ST Array 10574258 - -
Affymetrix Mouse Gene 1.0 ST Array 10574254 - -
Affymetrix Mouse Gene 1.0 ST Array 10574252 - -
Affymetrix Mouse Gene 1.0 ST Array 10574249 - -
Affymetrix Mouse Gene 1.0 ST Array 10574250 - -
Affymetrix Mouse Gene 1.0 ST Array 10574256 - -
Affymetrix Mouse Gene 1.0 ST Array 10574255 - -
Affymetrix Mouse Gene 1.0 ST Array 10574248 - -
Affymetrix Mouse Gene 1.0 ST Array 10574247 - -
Affymetrix Mouse Gene 1.0 ST Array 10574251 - -
Affymetrix HT MG-430 PM Array Plate 1436230_PM_at - -
Affymetrix Mouse Gene 1.1 ST Array 10574246 - -
Mouse Genome 430 2.0 Array 1436230_at - -
Mouse Genome 430 2.0 Array 1436230_PM_at - -
Affymetrix GeneChip Mouse Genome 430 2.0 Array 382045_at - -
Affymetrix GeneChip Mouse Genome 430 2.0 Array 382045_PM_at - -
Affymetrix Mouse Clariom S Array PSR0800008803.mm.1 NM_001033468.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0800008803.mm.1 NM_001033468.3 Refseq
Affymetrix Mouse Clariom S Array PSR0800008803.mm.1 NM_001145972.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0800008803.mm.1 NM_001145972.1 Refseq
Affymetrix Mouse Clariom S Array PSR0800008803.mm.1 TC0800001125.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0800008801.mm.1 NM_001033468.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0800008801.mm.1 NM_001033468.3 Refseq
Affymetrix Mouse Clariom S Array PSR0800008801.mm.1 NM_001145972.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0800008801.mm.1 NM_001145972.1 Refseq
Affymetrix Mouse Clariom S Array PSR0800008801.mm.1 TC0800001125.mm.1 MTA TC
Mouse Expression Set 430 1436230_PM_at - -
Mouse Expression Set 430 1436230_at - -
Affymetrix HT Mouse Genome 430B Array 1436230_at - -
Affymetrix HT Mouse Genome 430B Array 1436230_PM_at - -
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 865635 - -
Array Name Probe IDs Targets Target Type
Illumina mouse-6 v1.1 expression beadchip ILMN_1238976 - -
MouseRef-8 v2 Expression BeadChip ILMN_2856926 - -
Illumina MouseWG-6 v2.0 expression beadchip ILMN_2856926 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-l2rv2:80/100.66.78.26:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0