This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01247588_m1
See other ADGRG5 GE Assays ›
Gene Symbol:
Adgrg5
Gene Name:
adhesion G protein-coupled receptor G5
Gene Aliases:
Gm1109, Gpr114, PGR27
Chromosome Location:
Chr.8: 94921811 - 94943293 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01247588_m1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Adgrg5
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001033468.3 NP_001028640.2 9 - 10 1155 68
NM_001145972.1 NP_001139444.1 9 - 10 1163 68
GenBank mRNA AK028878.1 - 9 - 10 1156 68
AK156147.1 - 9 - 10 1274 68
AK156609.1 - 9 - 10 1274 68
BC144851.1 - 9 - 10 1124 9053399 68
BC145803.1 - 9 - 10 1124 40131106 68

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0800001125.mm.1

Set Membership:

Amplicon length less than or equal to 70 G-protein coupled receptor Mammalian Gene Collection Mus musculus Probe spans exons

Panther Classification:

Molecular Function -

G-protein coupled receptor

Gene Ontology Categories:

Function(s) Process(es)

signal transduction
cell surface receptor signaling pathway
G-protein coupled receptor signaling pathway
signal transducer activity
transmembrane signaling receptor activity
G-protein coupled receptor activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0800008799.mm.1 NM_001033468.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0800008799.mm.1 NM_001033468.3 Refseq
Affymetrix Mouse Clariom S Array PSR0800008799.mm.1 NM_001145972.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0800008799.mm.1 NM_001145972.1 Refseq
Affymetrix Mouse Clariom S Array PSR0800008799.mm.1 TC0800001125.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-l4hk6:80/100.66.76.55:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0