This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01247642_g1
See other TCF25 GE Assays ›
Gene Symbol:
Tcf25
Gene Name:
transcription factor 25 (basic helix-loop-helix)
Gene Aliases:
1100001J13Rik, 1810041K11Rik, D8Ertd325e, Nulp1, mKIAA1049
Chromosome Location:
Chr.8: 123373635 - 123404174 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01247642_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Tcf25
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001037877.3 NP_001032966.1 10 - 11 1282 74
NM_001037878.3 NP_001032967.1 10 - 11 1282 74
NM_001286362.1 NP_001273291.1 9 - 10 1207 74
NM_001286363.1 NP_001273292.1 9 - 10 1207 74
NM_025804.3 NP_080080.2 10 - 11 1282 74
GenBank mRNA AK003180.1 - 10 - 11 1170 74
AK013754.1 - 10 - 11 971 74
AK045397.1 - 9 - 10 1135 74
AK050274.1 - 10 - 11 1210 74
AK075728.1 - 10 - 11 1136 74
AK076151.1 - 4 - 5 369 74
AK088360.1 - 10 - 11 1170 74
AK089964.1 - 10 - 11 1171 74
AK122430.1 - 10 - 11 1137 74
AK148082.1 - 10 - 11 1144 74
AK150232.1 - 10 - 11 1169 74
AK150918.1 - 10 - 11 1163 74
AK150942.1 - 10 - 11 1156 74
AK151143.1 - 10 - 11 1156 74
AK151558.1 - 10 - 11 1163 74
AK152178.1 - 10 - 11 1156 74
AK152494.1 - 10 - 11 1156 74
AK161627.1 - 10 - 11 1158 74
AK167837.1 - 10 - 11 1156 74
AK168167.1 - 10 - 11 1156 74
BC016130.1 - 10 - 11 1144 74
BC025071.1 - 10 - 11 1143 3498003 74
BC046768.1 - 10 - 11 1240 5707312 74
EU624323.1 - 9 - 10 1045 74

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0800001520.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Tcf25

Gene Ontology Categories:

Function(s) Process(es)

negative regulation of transcription from RNA polymerase II promoter
transcription, DNA-templated
regulation of transcription, DNA-templated
DNA binding
transcription factor activity, sequence-specific DNA binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0800012460.mm.1 NM_001037877.3 Refseq
Affymetrix Mouse Clariom S Array PSR0800012460.mm.1 NM_001037878.3 Refseq
Affymetrix Mouse Clariom S Array PSR0800012460.mm.1 NM_001286362.1 Refseq
Affymetrix Mouse Clariom S Array PSR0800012460.mm.1 NM_001286363.1 Refseq
Affymetrix Mouse Clariom S Array PSR0800012460.mm.1 NM_025804.3 Refseq
Affymetrix Mouse Clariom S Array PSR0800012460.mm.1 TC0800001520.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-9hc65:80/100.66.78.230:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0